Product Name
Monoclonal Anti-B2M antibody produced in mouse clone 3F9-2C2, purified immunoglobulin, buffered aqueous solution
Product Number
WH0000567M1
Marque du produit
SIGMA
CAS Number
Poids moléculaire
Test | Caractéristique |
---|---|
Western Blotting (Cell lysate) | Antibody concentration: 1 ~ 5 ug/ml. Specific band of ~14 kDa using U-2 OS lysate. |
Immunohistochemistry-Paraffin Embedded Tissue (IHC-P) | Antibody concentration: 3 ug/ml using human esophagus tissue. |
Immunofluorescence (IF) | Antibody concentration: 10 ug/ml using HeLa cell. |
Sandwich ELISA | Detection limit for recombinant GST tagged B2M is approximately 0.03ng/ml. |
ELISA (Recombinant protein) | Antibody concentration: 1 ug/ml using recombinant protein. |
Western Blotting (Recombinant protein) | Antibody concentration: 1 ug/ml. Specific band of ~39 kDa using immunogen protein lysate. |