Skip to Content
Merck
Sign Into View Organizational & Contract Pricing

About This Item

Empirical Formula (Hill Notation):
C5H9NO
CAS Number:
Molecular Weight:
99.13
MDL number:
UNSPSC Code:
12352005
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

1-Methyl-2-pyrrolidone, for chromatography

grade

for chromatography

Quality Level

suitability

suitable for chromatography

SMILES string

N1(CCCC1=O)C

InChI

1S/C5H9NO/c1-6-4-2-3-5(6)7/h2-4H2,1H3

InChI key

SECXISVLQFMRJM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Methyl-2-pyrrolidone is a dipolar aprotic solvent widely used in organic chemistry for its ability to dissolve many chemicals. It is commonly used as a solvent or eluent for extracting soluble materials from soil samples using size-exclusion chromatography. NMP is a highly polar liquid and can be used for solvent extraction of polar substances from non-polar substances.

Application


  • Delivery of atovaquone and proguanil across sublingual membranes, in vitro.: This study explores the sublingual delivery of the antimalarial drugs atovaquone and proguanil, using 1-Methyl-2-pyrrolidone (NMP) as a solubilizing agent to enhance drug permeability. Results demonstrate NMP′s potential to improve drug delivery efficiency through sublingual membranes (Wallace et al., 2012).

  • Controlled delivery of aspirin: effect of aspirin on polymer degradation and in vitro release from PLGA based phase sensitive systems.: This research investigates the controlled release of aspirin from poly(lactic-co-glycolic acid) (PLGA) systems using NMP. The study highlights NMP′s role in modulating polymer degradation and drug release profiles, offering insights for developing controlled delivery systems (Tang and Singh, 2008).


Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

195.8 °F - Pensky-Martens closed cup

Flash Point(C)

91 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service