Skip to Content
Merck
All Photos(1)

Key Documents

71023-M

Sigma-Aldrich

TriEx Sf9 Cells - Novagen

Serum-free adapted Sf9 derivative for high-yield protein expression

Synonym(s):

Novagen® TriEx Sf9 Cells, Serum-free TriEx Sf9 Cells, TriEx Insect Cell, TriEx Sf9 Cells from Spodoptera frugiperda

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51111800
NACRES:
NA.54

Quality Level

storage condition

OK to freeze

shipped in

dry ice

storage temp.

≤ − 140°C

General description

The unique TriEx Sf9 Cells are derived from a high-yielding clone of Sf9 cells. Pre-adapted for growth in Novagen serum-free TriEx Insect Cell Medium, these cells are recommended for superior growth and protein yields by either baculovirus infection or transfection of appropriate vectors. For cotransfection of plasmids with linearized baculovirus DNA, we recommend Sf9 Insect Cells and BacVector Insect Cell Medium. After recovery from the frozen stock, the cells can be grown as semi-adherent cultures in tissue culture flasks or in suspension as shaker cultures. Cells are shipped on dry ice. Upon receipt, the vials should be removed from their container and either recovered immediately, or placed at -70°C and used within two weeks. For longer-term storage, place in liquid nitrogen.

Components

3 vials- Sf9 Insect Cells (frozen at 1 x 10 cells/ml)

Warning

Toxicity: Irritant

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service