Skip to Content
Merck
Sign Into View Organizational & Contract Pricing

About This Item

UNSPSC Code:
41171614
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

form

powder

Quality Level

shelf life

limited shelf life, expiry date on the label

application(s)

food and beverages
vitamins, nutraceuticals, and natural products

microbiology

storage temp.

2-8°C

suitability

Campylobacter spp.

General description

Park and Sanders Selective Supplement II, modified, is used as an antibiotic supplement in culture media to inhibit the growth of unwanted organisms while allowing the growth of the desired microorganisms. This antibiotic supplement is recommended for the selective isolation and enumeration of thermotolerant Campylobacter sp. The modified supplement consists of a combination of Amphotericin B and Cefoperazone with broad-spectrum antimicrobial activity against various Gram-positive and Gram-negative organisms.

Application

Park and Sanders Selective Supplement II, modified, is an antibiotic supplement that is recommended for the selective isolation and enumeration of thermotolerant Campylobacter species from various food and clinical samples.

Components

Composition
Per vial sufficient for 1000 ml medium
Cefoperazone 32mg
Amphotericin B 5mg

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service