grade
certified reference material
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
application(s)
microbiology
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
General description
This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Download your certificate at: http://www.sigma-aldrich.com.
Please specify if you require a single batch - in the remark field when ordering (only for orders with more than 15 packs)
Download your certificate at: http://www.sigma-aldrich.com.
Please specify if you require a single batch - in the remark field when ordering (only for orders with more than 15 packs)
Other Notes
Strain History< BCRC < ATCC, 1987 < G.W. Gorman WA-316-C3
Legal Information
Vitroids is a trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service