Skip to Content
Merck
Sign Into View Organizational & Contract Pricing

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~18 kDa

species reactivity

rat, mouse, human

concentration

1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:100
indirect immunofluorescence: 1:100-1:200
western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(S-T-T-P-G)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EIF4EBP1(1978)

General description

The gene EIF4EBP1 (eukaryotic translation initiation factor 4E binding protein 1) is mapped to human chromosome 8p12.

Immunogen

Peptide sequence around aa. 43-47 (S-T-T-P-G), according to the protein EIF4EBP1.

Biochem/physiol Actions

The gene EIF4EBP1 (eukaryotic translation initiation factor 4E binding protein 1) encodes a translation repressor that interacts with eukaryotic translation initiation factor 4E (eIF4E). eIF4E is a multisubunit complex that recruits 40S ribosomal subunits to the 5′ end of mRNA. The interaction of the encoded binding protein with this complex inhibits translation. The phosphorylation of eIF4Ebp1 in response to stimuli such as, UV irradiation and insulin signaling, results in dissociation of this factor from the eIF4E complex, leading to initiation of translation of mRNA. The encoded protein participates in cell proliferation, apoptosis, invasion, and metastasis. It functions as an effector molecule in mTOR (mammalian target of rapamycin complex 1) signaling pathway that regulates protein synthesis. It is found to be overexpressed in hepatocellular carcinoma tissues and serves as a potential prognostic and therapeutic target.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

4E-BP1 encodes one member of a family of translation repressor proteins. The protein directly interacts with eukaryotic translation initiation factor 4E (eIF4E), which is a limiting component of the multisubunit complex that recruits 40S ribosomal subunits to the 5' end of mRNAs. Interaction of this protein with eIF4E inhibits complex assembly and represses translation. This protein is phosphorylated in response to various signals including UV irradiation and insulin signaling, resulting in its dissociation from eIF4E and activation of mRNA translation.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service