Product Name
Diisobutylaluminum hydride solution 1.0 M in cyclohexane
Product Number
214949
Product Brand
ALDRICH
CAS Number
1191-15-7
Molecular Weight
142.22
Test | Specification |
---|---|
Appearance (Color) | Colorless |
Appearance (Form) | Liquid |
Hydrogen Evolution | 0,95 - 1,10 M |