Product Name
Tributyl citrate ≥97.0%
Product Number
27497
Product Brand
ALDRICH
CAS Number
77-94-1
Molecular Weight
360.44
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless |
APPEARANCE (FORM) | Liquid or Viscous Liquid |
PURITY (GC AREA %) | ≥ 97.0 % |
REFRACTIVE INDEX N20/D | 1.444 - 1.446 |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |