Product Name
3-Isochromanone 98%
Product Number
359351
Product Brand
ALDRICH
CAS Number
4385-35-7
Molecular Weight
148.16
Test | Specification |
---|---|
APPEARANCE | YELLOW TO BEIGE POWDER OR CRYSTALS WHICH MAY |
CONTAIN BROWN SPECS | |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE |
GAS LIQUID | 97.5% (MINIMUM) |
APPROVED SEPTEMBER 23, 2008 RJM |