Product Name
4-(Trifluoromethyl)phenylhydrazine 96%
Product Number
412295
Product Brand
ALDRICH
CAS Number
368-90-1
Molecular Weight
176.14
Test | Specification |
---|---|
Appearance (Color) | Conforms to Requirements |
Yellow to Orange to Brown | |
Appearance (Form) | Conforms to Requirement |
Powder, Crystals, Crystalline Powder and/or Chunks | |
Infrared spectrum | Conforms to Structure |
Carbon | 45.6 - 49.9 % |
Nitrogen | 15.2 - 16.6 % |
Purity (GC) | ≥95.5 % |