Product Name
Tetrabenzyl pyrophosphate 98%
Product Number
418633
Product Brand
ALDRICH
CAS Number
990-91-0
Molecular Weight
538.47
Test | Specification |
---|---|
APPEARANCE | WHITE TO OFF-WHITE POWDER OR CRYSTALS |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE. |
ELEMENTAL ANALYSIS | CARBON 60.9% - 63.4% |
HIGH PRESSURE LIQUID | 97.5% (MINIMUM) |
REVISED OCTOBER 16, 2007 RJM |