Product Name
Triisopropylsilanethiol 97%
Product Number
429937
Product Brand
ALDRICH
CAS Number
156275-96-6
Molecular Weight
190.42
Test | Specification |
---|---|
Appearance (Color) | Colorless to Light Yellow |
Appearance (Form) | Liquid |
Infrared spectrum | Conforms to Structure |
Purity (GC) | ≥96.5 % |