Product Name
Glutaconic acid 97% (T)
Product Number
49360
Product Brand
ALDRICH
CAS Number
1724-02-3
Molecular Weight
130.10
Test | Specification |
---|---|
APPEARANCE (COLOR) | White to Faint Beige |
APPEARANCE (FORM) | Powder or Crystals |
TITRATION (T) NAOH 0.1M | 96.5 - 103.5 % |
PURITY (HPLC AREA %) | ≥ 96.5 % |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |