Product Name
1,2-Dibromo-4,5-dimethylbenzene 97%
Product Number
638447
Product Brand
ALDRICH
CAS Number
24932-48-7
Molecular Weight
263.96
Test | Specification |
---|---|
APPEARANCE (COLOR) | White |
APPEARANCE (FORM) | Powder or Crystals or Flakes or Chunks |
TITRATION | 96.5 - 103.5 % |
TITRATION (METHOD) | WITH AGNO3 AFTER OXYGEN COMBUSTION |
PURITY (GC AREA %) | ≥ 96.5 % |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE |