Product Name
2,4-Dichloro-5-nitropyrimidine 97%
Product Number
658340
Product Brand
ALDRICH
CAS Number
49845-33-2
Molecular Weight
193.98
Test | Specification |
---|---|
APPEARANCE | WHITE TO BROWN SOLID OR LIQUID |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
TITRATION | 96.5% - 103.5% (WITH AGNO3 AFTER OXYGEN |
COMBUSTION) | |
AND/OR | |
ELEMENTAL ANALYSIS | CARBON 23.9% - 25.6% |
NITROGEN 20.9% - 22.4% | |
GAS LIQUID | 96.5% (MINIMUM) |
APPROVED JUNE 30, 2005 BTR |