Product Name
2-Amino-4-bromophenol ≥94%
Product Number
708356
Product Brand
ALDRICH
CAS Number
40925-68-6
Molecular Weight
188.02
Test | Specification |
---|---|
Appearance (Color) | White to Brown |
Appearance (Form) | Conforms to Requirements |
Powder or Crystals | |
Infrared spectrum | Conforms to Structure |
Carbon | 36.03 - 40.63 % |
Nitrogen | 7.00 - 7.89 % |
Purity (GC) | ≥94.0 % |