Product Name
Palladium on barium sulfate 10% Pd basis
Product Number
76020
Product Brand
ALDRICH
CAS Number
Molecular Weight
Test | Specification |
---|---|
APPEARANCE (COLOR) | Faint Brown to Dark Brown |
APPEARANCE (FORM) | Powder |
ASSAY (ATOMIC ABSORPT.) | 9 - 11 % PD |
ICP ANALYSIS | CONFIRMES BA, S AND PD |