Product Name
Hexyl salicylate ≥99.0% (GC)
Product Number
84280
Product Brand
ALDRICH
CAS Number
6259-76-3
Molecular Weight
222.28
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless |
APPEARANCE (FORM) | Liquid |
PURITY (GC AREA %) | ≥ 99.0 % |
REFRACTIVE INDEX N20/D | 1.504 - 1.506 |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE |
ACID VALUE | 0 - 5 |