Product Name
1-Butanol 99%
Product Number
B85900
Product Brand
ALDRICH
CAS Number
71-36-3
Molecular Weight
74.12
Test | Specification |
---|---|
APPEARANCE | COLORLESS LIQUID |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE AND STANDARD AS |
ILLUSTRATED ON PAGE 110A OF EDITION I, | |
VOLUME 1 OF "THE ALDRICH LIBRARY OF FT-IR | |
SPECTRA". | |
GAS LIQUID | 98.5% (MINIMUM) |
APPROVED MARCH 21, 1995 JSB |