Product Name
Cyanuric chloride 99%
Product Number
C95501
Product Brand
ALDRICH
CAS Number
108-77-0
Molecular Weight
184.41
Test | Specification |
---|---|
Appearance (Color) | White |
Appearance (Form) | Powder |
Infrared spectrum | Conforms to Structure |
Titration by AgNO3 | 98.5 - 101.5 % |
% Purity Based On Chloride Content By Titration | |
With AgNO3 After O2 Combustion | |
Titration by AgNO3 | 98.5 - 101.5 % |
% Purity By Titration With AgNO3 After Reflux | |
Solubility (Turbidity) | Clear to Slightly Hazy |
Solubility (Color) | Colorless to Pale Yellow |
50mg/ml, Acetone |