Product Name
2,6-Dimethylbenzoquinone 99%
Product Number
D149705
Product Brand
ALDRICH
CAS Number
527-61-7
Molecular Weight
136.15
Test | Specification |
---|---|
APPEARANCE | YELLOW TO YELLOW-ORANGE TO GOLD FIBERS OR |
POWDER | |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE AND STANDARD AS |
ILLUSTRATED ON PAGE 452D OF EDITION I, | |
VOLUME 1 OF "THE ALDRICH LIBRARY OF FT-IR | |
SPECTRA". | |
GAS LIQUID | 98.5% (MINIMUM) |
APPROVED JANUARY 30, 1995 JSB |