Product Name
2′-Methoxyacetophenone 99%
Product Number
M9203
Product Brand
ALDRICH
CAS Number
579-74-8
Molecular Weight
150.17
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless |
APPEARANCE (FORM) | Liquid |
PURITY (GC AREA %) | ≥ 98.5 % |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE |