Product Name
N-Phenylthiourea ≥98%
Product Number
P7629
Product Brand
ALDRICH
CAS Number
103-85-5
Molecular Weight
152.22
Test | Specification |
---|---|
APPEARANCE (COLOR) | White to Faint Brown |
APPEARANCE (FORM) | Powder |
PURITY (HPLC AREA %) | ≥ 97.5 % |
SOLUBILITY (COLOR) | Colorless to Very Faint Yellow |
SOLUBILITY (TURBIDITY) | Clear to Hazy |
SOLUBILITY (METHOD) | 50MG/ML ETHANOL |