Product Name
Trichloroisocyanuric acid for synthesis
Product Number
8.08607
Product Brand
MM
CAS Number
87-90-1
Molecular Weight
232.41
Test | Specification |
---|---|
REDOX TITRATION | 98.0 - 102.0 % |
REDOXTITRATION (METHOD) | Iodometric |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE |