Product Name
Synperonic® F 108 surfactant, non-ionic
Product Number
07579
Product Brand
SIAL
CAS Number
9003-11-6
Molecular Weight
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless or White |
APPEARANCE (FORM) | Crystals or Flakes |
MELTING POINT | 57 - 61 C |
WATER | ≤ 0.5 % |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE |
HYDROXYL VALUE | 6 - 9 mg KOH/g |