Product Name
4-Isopropylthioxanthone PESTANAL®, analytical standard
Product Number
34222
Product Brand
SIAL
CAS Number
83846-86-0
Molecular Weight
254.35
Test | Specification |
---|---|
APPEARANCE (COLOR) | Faint Yellow to Very Dark Yellow and Faint Beige to Beige and Faint Brown to Brown and Yellow-Beige and Yellow-Brown and Yellow-Green |
APPEARANCE (FORM) | Powder or Crystals |
PURITY (HPLC AREA %) | ≥ 99.0 % |
MELTING POINT | FOR INFORMATION ONLY |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
RESIDUAL SOLVENTS | ≤ 1.0 % |
EXPIRY PERIOD | 60 MONTHS |