Product Name
Bis(2-ethylhexyl) phthalate PESTANAL®, analytical standard
Product Number
36735
Product Brand
SIAL
CAS Number
117-81-7
Molecular Weight
390.56
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless to Light Yellow |
APPEARANCE (FORM) | Liquid |
PURITY (GC AREA %) | ≥ 98.0 % |
REFRACTIVE INDEX N20/D | 1.4846 - 1.4886 |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
EXPIRY PERIOD | 60 MONTHS |