Product Name
4-Phenyl-1,2,4-triazoline-3,5-dione derivatization grade (HPLC), LiChropur™, ≥98.0% (CHN)
Product Number
42579
Product Brand
SIAL
CAS Number
4233-33-4
Molecular Weight
175.14
Test | Specification |
---|---|
APPEARANCE (COLOR) | Orange and Red to Dark Red and Orange-Red and Red-Orange |
APPEARANCE (FORM) | Powder or Crystals |
ASSAY (ELEMENTAL ANALYSIS) | ≥ 98.0 |
CARBON CONTENT | 54.86 % (theory) |
NITROGEN CONTENT | 23.99 % (theory) |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |