Product Name
Amitrol PESTANAL®, analytical standard
Product Number
45324
Product Brand
SIAL
CAS Number
61-82-5
Molecular Weight
84.08
Test | Specification |
---|---|
APPEARANCE (COLOR) | White to Light Yellow |
APPEARANCE (FORM) | Powder or Crystals |
PURITY (GC AREA %) | ≥ 98.0 % |
IDENTIFICATION (GC-MS) | IDENTITY CORRESPONDS |
MELTING POINT | 154 - 159 C |
WATER | ≤ 1.0 % |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
EXPIRY PERIOD | 60 MONTHS |