Product Name
Chloralose PESTANAL®, analytical standard
Product Number
45373
Product Brand
SIAL
CAS Number
15879-93-3
Molecular Weight
309.53
Test | Specification |
---|---|
APPEARANCE (COLOR) | White to Off White |
APPEARANCE (FORM) | Powder or Crystals |
PURITY (HPLC AREA %) | ≥ 85% Alpha-Isomers |
HPLC MINOR COMPONENT (AREA %) | ≤ 15% beta-Isomers |
MELTING POINT | 180 - 188 C |
WATER | ≤ 1.0 % |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
EXPIRY PERIOD | 60 MONTHS |