Product Name
Nitenpyram PESTANAL®, analytical standard
Product Number
46077
Product Brand
SIAL
CAS Number
150824-47-8
Molecular Weight
270.72
Test | Specification |
---|---|
APPEARANCE (COLOR) | White to Very Dark Yellow to Very Dark Beige |
APPEARANCE (FORM) | Powder or Crystals |
PURITY (HPLC AREA %) | ≥ 98.0 % |
MELTING POINT | 79 - 85 C |
WATER | ≤ 1.0 % |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
EXPIRY PERIOD | 60 MONTHS |