Product Name
Cetyl alcohol analytical standard
Product Number
68824
Product Brand
SIAL
CAS Number
36653-82-4
Molecular Weight
242.44
Test | Specification |
---|---|
APPEARANCE (COLOR) | White |
APPEARANCE (FORM) | Powder or Crystals or Flakes or Pellets |
PURITY (GC AREA %) | ≥ 99.5 % |
MELTING POINT | 48 - 50 C |
SOLUBILITY (COLOR) | Colorless |
SOLUBILITY (TURBIDITY) | Clear |
SOLUBILITY (METHOD) | 1G IN 10ML ETHANOL |
CARBON CONTENT | 79.26 % (THEORY) |
HYDROGEN CONTENT | 14.13 % (THEORY) |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
EXPIRY PERIOD | 36 MONTHS |