Product Name
Silylating mixture II according to Horning LiChropur™
Product Number
85432
Product Brand
SIAL
CAS Number
101660-05-3
Molecular Weight
Test | Specification |
---|---|
APPEARANCE (COLOR) | Faint Yellow to Yellow |
APPEARANCE (FORM) | Liquid |
REFRACTIVE INDEX N20/D | 1.434 - 1.438 |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |