Product Name
Isobutyryl-L-carnitine analytical standard
Product Number
91299
Product Brand
SIAL
CAS Number
25518-49-4
Molecular Weight
231.29
Test | Specification |
---|---|
APPEARANCE (COLOR) | White to Off White |
APPEARANCE (FORM) | Powder or Crystals |
PURITY (HPLC AREA %) | ≥ 97.0 % |
SPECIFIC ROTATION (20/D) | -24.0 - -20.0 Degrees |
CONCENTRATION | c=0.1 in water |
WATER | ≤ 10.0 % Water (Calculated from Elemental Analysis) |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
EXPIRY PERIOD | 36 MONTHS |