Product Name
Tricosane analytical standard
Product Number
91447
Product Brand
SIAL
CAS Number
638-67-5
Molecular Weight
324.63
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless or White |
APPEARANCE (FORM) | Powder or Crystals or Flakes |
PURITY (GC AREA %) | ≥ 99.5 % |
MELTING POINT | 47 - 49 C |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
EXPIRY PERIOD | 36 MONTHS |