Product Name
Calcium phosphate dibasic dihydrate puriss., meets analytical specification of Ph. Eur., BP, USP, 98-102.5%
Product Number
04231
Product Brand
SIGALD
CAS Number
7789-77-7
Molecular Weight
172.09
Test | Specification |
---|---|
APPEARANCE (COLOR) | White to Off White |
APPEARANCE (FORM) | Powder |
ASSAY | 98 - 102.5 % (USP) |
LOSS ON IGNITION | 24.5-26.5% 800 C |
TRACE ANALYSIS (ADDITIONAL TESTS) | Ba: in accordance (USP, PH. EUR)-br-CARBONATE (CO3) COMPLYING-br-heavy metals (as Pb) ≤ 0.003 % |
IRON (ICP) | ≤ 300 mg/kg |
ARSENIC TRACES (MHS-AAS) | ≤ 1 mg/kg |
CHLORIDE (CL) | ≤ 200 mg/kg |
SULFATE (SO4) | ≤ 3000 mg/kg |
Substances Insoluble in HCl | ≤ 1000 mg/kg |
REMARKS | ORGANIC VOLATILE IMPURITIES COMPLYING RESIDUAL SOLVENTS COMPLYING-br-FLUORIDE (F) ≤ 50 MG/KG |
EXPIRY PERIOD | 72 MONTHS |