Product Name
Q-Dextran average mol wt 4,000
Product Number
19534
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
APPEARANCE (COLOR) | White to Off White |
APPEARANCE (FORM) | Powder or Crystals or Granules |
MOLECULAR NUMBER | NUMBER AVERAGE MOLECULAR WEIGHT (MN) REPORT RESULT |
MOLECULAR WEIGHT DETERMINATION | WEIGHT AVERAGE MOLECULAR WEIGHT (MW) 3500 - 5500 |
LOSS ON DRYING | ≤ 10 % |
NITROGEN CONTENT | 1.5 - 2.5 % |