Product Name
Guanosine 5′-diphosphate disodium salt ≥90% (HPLC)
Product Number
51060
Product Brand
SIGMA
CAS Number
7415-69-2
Molecular Weight
487.16
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless or White |
APPEARANCE (FORM) | Powder |
PURITY (HPLC AREA %) | ≥ 90.0 % |
SOLUBILITY (COLOR) | Colorless |
SOLUBILITY (TURBIDITY) | Clear |
SOLUBILITY (METHOD) | 50 MG/ML IN WATER |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE |
RECOMMENDED RETEST PERIOD | 36 MONTHS |