Product Name
7α-Hydroxy-4-cholesten-3-one ≥95.0% (HPLC)
Product Number
52853
Product Brand
SIGMA
CAS Number
3862-25-7
Molecular Weight
400.64
Test | Specification |
---|---|
APPEARANCE (COLOR) | White to Off White |
APPEARANCE (FORM) | Powder or Crystals |
PURITY (HPLC AREA %) | ≥ 95.0 % |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE |