Product Name
7-Methoxycoumarin suitable for fluorescence, ≥98.0% (TLC)
Product Number
64951
Product Brand
SIGMA
CAS Number
531-59-9
Molecular Weight
176.17
Test | Specification |
---|---|
APPEARANCE (COLOR) | White to Faint Brown |
APPEARANCE (FORM) | Crystals |
PURITY (TLC AREA %) | ≥ 98.0 % |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
EMISSION MAXIMUM | 385 ± 5 nm |
REMARKS ON LUMINESCENCE | Exc.: 350 nm in CHCl3 |