Product Name
N-(3-Hydroxydecanoyl)-DL-homoserine lactone
Product Number
68873
Product Brand
SIGMA
CAS Number
929222-14-0
Molecular Weight
271.35
Test | Specification |
---|---|
APPEARANCE (COLOR) | White |
APPEARANCE (FORM) | Powder or Crystals |
PURITY (HPLC AREA %) | ≥ 95.0 % |
CARBON CONTENT | 61.97 (theory) |
NITROGEN CONTENT | 5.16 (theory) |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
RECOMMENDED RETEST PERIOD | 36 MONTHS |