Product Name
Uricase from Bacillus fastidiosus lyophilized, ~9 U/mg
Product Number
94310
Product Brand
SIGMA
CAS Number
9002-12-4
Molecular Weight
Test | Specification |
---|---|
APPEARANCE (COLOR) | White to Faint Beige |
APPEARANCE (FORM) | Powder |
SOLUBILITY (COLOR) | Colorless to Very Faint Yellow |
SOLUBILITY (TURBIDITY) | Clear |
SOLUBILITY (METHOD) | 5 mg/ml in enzyme dilution buffer |
ENZYMATIC ACTIVITY | ≥ 7.2 U/mg |
RECOMMENDED RETEST PERIOD | 54 MONTHS |