Product Name
Z-Arg-Arg-7-amido-4-methylcoumarin hydrochloride
Product Number
C5429
Product Brand
SIGMA
CAS Number
136132-67-7
Molecular Weight
621.69
Test | Specification |
---|---|
APPEARANCE | WHITE POWDER |
SOLUBILITY | CLEAR COLORLESS SOLUTION AT 50 MG/ML IN METHANOL |
WATER CONTENT BY KARL FISCHER | REPORT RESULT |
CARBON | 53.5 - 58.0% * |
NITROGEN | 18.7 - 20.5% * |
CHLORIDE | > OR = 5.5% |
PURITY BY THIN LAYER CHROMATOGRAPHY | > OR = 98% |
NOTE: FORMULA AND FORMULA WEIGHT ARE FOR THE FREE ACID | |
*CORRECTED FOR WATER AND SALT CONTENT | |
DOCUMENT # | C5429/05/09/08/1 |