Product Name
Dulbecco’s Modified Eagle’s Medium - low glucose With 1000 mg/L glucose, and sodium bicarbonate, without L-glutamine and phenol red, liquid, sterile-filtered, suitable for cell culture
Product Number
D5921
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
ORIGIN | THIS IS AN ANIMAL DERIVED COMPONENT FREE PRODUCT |
APPEARANCE | CLEAR SOLUTION |
PH TEST AT RT | 7.0 - 7.6 |
OSMOLALITY TEST | 289 - 320 MOSM/KG H2O |
STERILE BY USP | STERILE |
ENDOTOXIN ASSAY | NMT 1 EU/ML |
GLUCOSE | 0.9 - 1.1 G/L |
KEY ELEMENTS BY ICP | CONSISTENT WITH FORMULA |
AMINO ACIDS BY HPLC | CONSISTENT WITH FORMULA |
CELL CULTURE TEST | PASS |
EXPIRATION DATE | 18 MONTHS |
D5921:08/03-2 |