Product Name
Poly-L-lysine solution 0.01%, sterile-filtered, BioReagent, suitable for cell culture
Product Number
P4707
Product Brand
SIGMA
CAS Number
25988-63-0
Molecular Weight
Test | Specification |
---|---|
APPEARANCE | CLEAR TO CLOUDY COLORLESS LIQUID |
STERILE BY USP XXIII | STERILE |
PERFORMANCE TEST | PASS ATTACHMENT ASSAY |
CELL CULTURE TEST | PASS TOXICITY ASSAY |
LYSINE CONTENT | NLT 0.45 MM |
ENDOTOXIN ASSAY | REPORT RESULTS |
EXPIRATION DATE | 24 MONTHS |
DOCUMENT # P4707/02/10/00/6 |