Product Name
Rhodamine 6G Dye content ~95 %
Product Number
R4127
Product Brand
SIGMA
CAS Number
989-38-8
Molecular Weight
479.01
Test | Specification |
---|---|
APPEARANCE | BROWN-RED POWDER |
SOLUBILITY | DARK RED SOLUTION AT 1MG/ML IN WATER |
ELEMENTAL ANALYSIS | 66.5 TO 74.0% CARBON |
5.0 TO 6.2% NITROGEN | |
DYE CONTENT | APPROX. 95% |
ULTRAVIOLET/VISIBLE SPECTRUM | E1% = GREATER THAN 1700 AT LAMBDA MAX APPROX. 525NM IN WATER |
APPROVED JLH 5/9/94 |