Product Name
Saccharin sodium salt hydrate ≥98.0% (titration)
Product Number
S1002
Product Brand
SIGMA
CAS Number
82385-42-0
Molecular Weight
205.17 (anhydrous basis)
Test | Specification |
---|---|
APPEARANCE | WHITE TO OFF-WHITE POWDER OR PELLETS |
SOLUBILITY | CLEAR COLORLESS SOLUTION AT 100MG/ML IN WATER |
MOISTURE CONTENT | 4.0% TO 15.0% |
PURITY BY TITRATION * | > OR = 98.0% |
PURITY BY THIN LAYER CHROMATOGRAPHY | > OR = 99% |
* ANHYDROUS BASIS | |
DOCUMENT # | S1002/11/17/08/6 |