Product Name
ANTI-GUCY2C antibody produced in mouse clone 1E1, purified immunoglobulin, buffered aqueous solution
Product Number
SAB1412659
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Appearance | colorless liquid |
Concentration | Report Result |
Storage/Stability | Report Result |
Storage Temperature | -20�C |
Species Reactivity | Report Result |
ELISA | Antibody concentration: 1 ug/ml using recombinant protein. |
Sandwich ELISA | Detection limit for recombinant GST tagged GUCY2C is approximately 0.3ng/ml. |
Western Blotting (Recombinant Protein) | Antibody concentration: 1 ug/ml. Specific band of ~37.7 kDa using immunogen protein lysate. |
Western Blotting (Cell Lysate) | Antibody concentration: 1 ~ 5 ug/ml. Specific band of ~123 kDa using COLO 320 HSR lysate. |
Western Blotting (Tissue lysate) | Not Verified |
Western Blotting (Transfected lysate) | Not Verified |
Immunohistochemistry | Not Verified |
Immunofluorescence | Not Verified |
Handling | Report Result |