Product Name
Anti-PIWIL2, (N-terminal) antibody produced in rabbit affinity isolated antibody
Product Number
SAB2105190
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Concentration | 0.5-1 mg/ml |
ELISA | Not Verified |
Western Blotting | Antibody dilution: 1 µg/ml |
Immunohistochemistry | Not Verified |
Immunofluorescence | Not Verified |
Immunoprecipitation | Not Verified |
Flow Cytometry | Not Verified |
Chromatin Immunoprecipitation | Not Verified |