Product Name
Anti-SOX2 antibody produced in rabbit affinity isolated antibody, buffered aqueous solution
Product Number
SAB3500363
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Concentration | 1 mg/mL |
ELISA | 0.01-1 ug/mL |
Western Blotting | 1-2 ug/mL |
Immunohistochemistry | Not Verified |
Immunocytochemistry | Not Verified |
Immunofluorescence | Not Verified |
Immunoprecipitation | Not Verified |
Flow Cytometry | Not Verified |
Chromatin Immunoprecipitation | Not Verified |